accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)OHOOHOO(E)

CAS RN 17924-92-4

IUPAC Name: (3S,11E)-14,16-dihydroxy-3-methyl-3,4,5,6,7,8,9,10-octahydro-1H-2-benzoxacyclotetradecine-1,7-dione
Synonyms: (-)-zearalenone F-2 toxin (S)-(-)-zearalenone trans-zearalenone (S)-zearalenone FES zearalenone F 2 toxin mycotoxin F2 ccris 623
Molecular Weight (g/mol): 318.37
Molecular Formula: C18H22O5
InChi Key: MBMQEIFVQACCCH-QBODLPLBSA-N
SMILES: C[C@H]1CCCC(=O)CCC\C=C\C2=CC(O)=CC(O)=C2C(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results