accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17933-03-8

IUPAC Name: (3-methylphenyl)boronic acid
Synonyms: 3-methylphenylboronic acid
Molecular Weight (g/mol): 135.96
Molecular Formula: C7H9BO2
InChi Key: BJQCPCFFYBKRLM-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (7)
  • (1)
Quantity 
  • (3)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (1)
  • (7)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)