Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M13145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1314-5G 5 g
M1314-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1314-5G Supplier TCI America Supplier No. M13145G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17933-03-8
Molecular Formula C7H9BO2
MDL Number MFCD00040198
InChI Key BJQCPCFFYBKRLM-UHFFFAOYSA-N
Synonym 3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid
PubChem CID 2733950
IUPAC Name (3-methylphenyl)boronic acid

Specifications

Quantity 5 g
SMILES CC1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 135.96
Formula Weight 135.96
Physical Form Crystalline Powder
RTECSNumber ED7788888
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.