accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 179334-18-0

IUPAC Name: 1-[(2,5-difluorophenyl)methyl]piperazine
Synonyms: 1-[(2,5-difluorophenyl)methyl]piperazine
Molecular Weight (g/mol): 212.24
Molecular Formula: C11H14F2N2
InChi Key: VTKXPESKKMTHJP-UHFFFAOYSA-N
SMILES: FC1=CC(CN2CCNCC2)=C(F)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results