accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNNHONOOHOOH(E/Z)

CAS RN 179386-44-8

IUPAC Name: 10-(methylamino)-1,3-diazatricyclo[6.3.1.0⁴,¹²]dodeca-4,6,8(12)-trien-2-one; but-2-enedioic acid
Synonyms: butenedioic acid; sumanirole
Molecular Weight (g/mol): 319.32
Molecular Formula: C15H17N3O5
InChi Key: VOJRMYBBPKNLLI-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CNC1CN2C(=O)NC3=CC=CC(C1)=C23

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)