accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1794-89-4

H3CBrBrBrBrOHO

CAS RN 1794-89-4

IUPAC Name: 9,10,12,13-tetrabromooctadecanoic acid
Synonyms: 9,10,12,13-tetrabromooctadecanoic acid
Molecular Weight (g/mol): 600.07
Molecular Formula: C18H32Br4O2
InChi Key: HTORNZPNCYXGMR-UHFFFAOYNA-N
SMILES: CCCCCC(Br)C(Br)CC(Br)C(Br)CCCCCCCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results