accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1796-92-5

IUPAC Name: disodium 5-{[(1E)-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene](2,6-dichlorophenyl)methyl}-2-hydroxy-3-methylbenzoate
Synonyms: disodium 5-{[(1E)-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene](2,6-dichlorophenyl)methyl}-2-hydroxy-3-methylbenzoate
Molecular Weight (g/mol): 503.24
Molecular Formula: C23H14Cl2Na2O6
InChi Key: JROAZQFKSSYEBL-YWABOPJLSA-L
SMILES: [Na+].[Na+].CC1=CC(=CC(C([O-])=O)=C1O)C(=C1\C=C(C)C(=O)C(=C1)C([O-])=O)\C1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results