Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chromazurol B

Catalog No. AC464530250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464530250 25 g
AC464530050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464530250 Supplier Thermo Scientific Chemicals Supplier No. 464530250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
 Restricted Item
Add to Cart

CAS: 1796-92-5 | C23H14Cl2Na2O6 | 503.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1796-92-5
Molecular Formula C23H14Cl2Na2O6
Molecular Weight (g/mol) 503.24
InChI Key JROAZQFKSSYEBL-YWABOPJLSA-L
IUPAC Name disodium 5-{[(1E)-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene](2,6-dichlorophenyl)methyl}-2-hydroxy-3-methylbenzoate
SMILES [Na+].[Na+].CC1=CC(=CC(C([O-])=O)=C1O)C(=C1\C=C(C)C(=O)C(=C1)C([O-])=O)\C1=C(Cl)C=CC=C1Cl

Specifications

Quantity 25 g
Formula Weight 503.25
Chemical Name or Material Chromazurol B
EINECSNumber 217-279-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.