Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrome Pure Blue BX, TCI America™
SDP

Catalog No. C033725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C033725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C033725G Supplier TCI America Supplier No. C033725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1796-92-5
Molecular Formula C23H14Cl2Na2O6
Molecular Weight (g/mol) 503.239
MDL Number MFCD00036446
InChI Key JROAZQFKSSYEBL-UHFFFAOYSA-L
Synonym Eriochrome Azurol B, Sunchromine Pure Blue BX, Mordant Blue 1
PubChem CID 54726806
IUPAC Name disodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichlorophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC=C3Cl)Cl.[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 503.24
Physical Form Crystalline Powder
Chemical Name or Material Chrome Pure Blue BX
EINECSNumber (5)-2082
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.