accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18022-66-7

IUPAC Name: 3,4-dimethylcyclohex-3-ene-1-carbaldehyde
Synonyms: 3,4-dimethylcyclohex-3-ene-1-carbaldehyde
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: NOJNKSAQZMVMFF-UHFFFAOYNA-N
SMILES: CC1=C(C)CC(CC1)C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results