accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3OH3C

CAS RN 18066-68-7

IUPAC Name: ethyl 2-(3,4-dimethoxyphenyl)acetate
Synonyms: ethyl 2-(3,4-dimethoxyphenyl)acetate
Molecular Weight (g/mol): 224.26
Molecular Formula: C12H16O4
InChi Key: WZKCZNJTDZCNMH-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(OC)C(OC)=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results