accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CH3COOOOHNHOO

CAS RN 180675-08-5

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-[(3-methylpentan-3-yl)oxy]-4-oxobutanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-[(3-methylpentan-3-yl)oxy]-4-oxobutanoic acid
Molecular Weight (g/mol): 439.51
Molecular Formula: C25H29NO6
InChi Key: SYGKUYKLHYQKPL-UHFFFAOYNA-N
SMILES: CCC(C)(CC)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)