accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18084-64-5

IUPAC Name: 1,4,7,10-tetrabenzyl-1,4,7,10-tetraazacyclododecane
Synonyms: 1,4,7,10-tetrabenzyl-1,4,7,10-tetraazacyclododecane
Molecular Weight (g/mol): 532.78
Molecular Formula: C36H44N4
InChi Key: VXOJKUWHCFFUCO-UHFFFAOYSA-N
SMILES: C(N1CCN(CC2=CC=CC=C2)CCN(CC2=CC=CC=C2)CCN(CC2=CC=CC=C2)CC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results