Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,7,10-Tetrabenzyl-1,4,7,10-tetraazacyclododecane 97.0+%, TCI America™
SDP

Catalog No. T33565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33565G 5 g
T33561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33565G Supplier TCI America Supplier No. T33565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18084-64-5
Molecular Formula C36H44N4
Molecular Weight (g/mol) 532.776
InChI Key VXOJKUWHCFFUCO-UHFFFAOYSA-N
PubChem CID 428363
IUPAC Name 1,4,7,10-tetrabenzyl-1,4,7,10-tetrazacyclododecane
SMILES C1CN(CCN(CCN(CCN1CC2=CC=CC=C2)CC3=CC=CC=C3)CC4=CC=CC=C4)CC5=CC=CC=C5

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 5 g
Formula Weight 532.78
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,7,10-Tetrabenzyl-1,4,7,10-tetraazacyclododecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.