accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOH3CNHPOOONNNNNH2OHHOO

CAS RN 1809249-37-3

IUPAC Name: 2-ethylbutyl 2-({[(5-{4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl}-5-cyano-3,4-dihydroxyoxolan-2-yl)methoxy](phenoxy)phosphoryl}amino)propanoate
Synonyms: 2-ethylbutyl 2-{[(5-{4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl}-5-cyano-3,4-dihydroxyoxolan-2-yl)methoxy(phenoxy)phosphoryl]amino}propanoate
Molecular Weight (g/mol): 602.59
Molecular Formula: C27H35N6O8P
InChi Key: RWWYLEGWBNMMLJ-UHFFFAOYNA-N
SMILES: CCC(CC)COC(=O)C(C)NP(=O)(OCC1OC(C#N)(C(O)C1O)C1=CC=C2N1N=CN=C2N)OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)