CAS RN 181217-39-0
IUPAC Name:
6-chloro-1-(4-hydroxyphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrochloride
Synonyms:
fenoldopam hydrochloride
Molecular Weight (g/mol):
342.22
Molecular Formula:
C16H17Cl2NO3
InChi Key:
NLMPGIXLXSPNFS-UHFFFAOYNA-N
SMILES:
Cl.OC1=CC=C(C=C1)C1CNCCC2=C(Cl)C(O)=C(O)C=C12
1
–
1
of
1
results
1
R&D Systems™ Fenoldopam hydrochloride
CAS: 181217-39-0 Molecular Formula: C16H17Cl2NO3