accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHO

CAS RN 181308-56-5

IUPAC Name: tert-butyl N-(4-formylcyclohexyl)carbamate
Synonyms: tert-butyl N-(4-formylcyclohexyl)carbamate
Molecular Weight (g/mol): 227.3
Molecular Formula: C12H21NO3
InChi Key: GPDBIGSFXXKWQR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CC1)C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results