accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3CH3Cl

CAS RN 18162-84-0

IUPAC Name: chlorodimethyloctylsilane
Synonyms: chlorodimethyloctylsilane
Molecular Weight (g/mol): 206.83
Molecular Formula: C10H23ClSi
InChi Key: DBKNGKYVNBJWHL-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si](C)(C)Cl
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)