Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodimethyl-n-octylsilane, 97%

Catalog No. AAL1656406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1656406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1656406 Supplier Thermo Scientific Chemicals Supplier No. L1656406

missing translation for 'validMainframeMsg'

CAS: 18162-84-0 | C10H23ClSi | 206.829 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18162-84-0
Molecular Formula C10H23ClSi
Molecular Weight (g/mol) 206.829
MDL Number MFCD00000511
InChI Key DBKNGKYVNBJWHL-UHFFFAOYSA-N
Synonym n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc
PubChem CID 87482
IUPAC Name chloro-dimethyl-octylsilane
SMILES CCCCCCCC[Si](C)(C)Cl

Specifications

Boiling Point 222°C to 225°C
Quantity 5 g
UN Number UN2987
Beilstein 1738376
Flash Point 92°C (198°F)
Formula Weight 206.83
Percent Purity 97%
Odor Acrid
Refractive Index 1.435
Sensitivity Moisture sensitive
Density 0.873
Chemical Name or Material Chlorodimethyl-n-octylsilane
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 242-044-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.