accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNHNNN

CAS RN 18200-13-0

IUPAC Name: N-[2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]-5,6,7,8-tetrahydronaphthalen-1-amine
Synonyms: N-[2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]-5,6,7,8-tetrahydronaphthalen-1-amine
Molecular Weight (g/mol): 291.36
Molecular Formula: C17H17N5
InChi Key: YYNRZIFBTOUICE-UHFFFAOYSA-N
SMILES: C1CCC2=C(C1)C=CC=C2NC1=CC=CC=C1C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)