accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 182250-70-0

CH3H3CCH3SiOOHOSiH3CCH3H3C

CAS RN 182250-70-0

IUPAC Name: {3,5-bis[(tert-butyldiphenylsilyl)oxy]phenyl}methanol
Synonyms: {3,5-bis[(tert-butyldiphenylsilyl)oxy]phenyl}methanol
Molecular Weight (g/mol): 616.95
Molecular Formula: C39H44O3Si2
InChi Key: SDDISUZNVYXXHS-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](OC1=CC(O[Si](C2=CC=CC=C2)(C2=CC=CC=C2)C(C)(C)C)=CC(CO)=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results