accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHNHNONHNHOOCH3CH3H3C

CAS RN 1822851-77-3

IUPAC Name: tert-butyl N-(3-{2-[(3-{[(tert-butoxy)carbonyl]amino}propyl)amino]acetamido}propyl)carbamate
Synonyms: tert-butyl N-{3-[2-({3-[(tert-butoxycarbonyl)amino]propyl}amino)acetamido]propyl}carbamate
Molecular Weight (g/mol): 388.51
Molecular Formula: C18H36N4O5
InChi Key: LUUNBXYJZAFAKK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCNCC(=O)NCCCNC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results