accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PONHOH

CAS RN 182485-36-5

IUPAC Name: methyl(1,2,3,6-tetrahydropyridin-4-yl)phosphinic acid
Synonyms: methyl(1,2,3,6-tetrahydropyridin-4-yl)phosphinic acid
Molecular Weight (g/mol): 161.14
Molecular Formula: C6H12NO2P
InChi Key: MFUKVPOVVKKLRQ-UHFFFAOYNA-N
SMILES: CP(O)(=O)C1=CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results