CAS RN 1825-21-4
CAS RN 1825-21-4
IUPAC Name:
1,2,3,4,5-pentachloro-6-methoxybenzene
Synonyms:
pentachloroanisole
Molecular Weight (g/mol):
280.35
Molecular Formula:
C7H3Cl5O
InChi Key:
BBABSCYTNHOKOG-UHFFFAOYSA-N
SMILES:
COC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl
1
–
2
of
2
results
1
2,3,4,5,6-Pentachloroanisole, SPEX CertiPrep™

CAS: 1825-21-4 Molecular Formula: C7H3Cl5O Molecular Weight (g/mol): 280.35 InChI Key: BBABSCYTNHOKOG-UHFFFAOYSA-N PubChem CID: 15767 IUPAC Name: 1,2,3,4,5-pentachloro-6-methoxybenzene SMILES: COC1=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl