Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentachloroanisole, Restek
SDP

Catalog No. 06719887
Click to view available options
Quantity:
1 mL

Chemical Identifiers

CAS 1825-21-4
Molecular Formula C7H3Cl5O
Molecular Weight (g/mol) 280.35
InChI Key BBABSCYTNHOKOG-UHFFFAOYSA-N
PubChem CID 15767
IUPAC Name 1,2,3,4,5-pentachloro-6-methoxybenzene
SMILES COC1=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl

Specifications

Quantity 1 mL
Chemical Name or Material Pentachloroanisole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.