accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SN

CAS RN 1826-16-0

IUPAC Name: 2-methyl-4-phenyl-1,3-thiazole
Synonyms: 2-methyl-4-phenyl-1,3-thiazole
Molecular Weight (g/mol): 175.25
Molecular Formula: C10H9NS
InChi Key: XROORURTAQOYLW-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results