accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2MgBr

CAS RN 1826-67-1

IUPAC Name: bromo(ethenyl)magnesium
Synonyms: bromo(ethenyl)magnesium
Molecular Weight (g/mol): 131.26
Molecular Formula: C2H3BrMg
InChi Key: XHHHAXOHMKAOSL-UHFFFAOYSA-M
SMILES: Br[Mg]C=C
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results