accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 183364-82-1

POOHHOOOHH2N

CAS RN 183364-82-1

IUPAC Name: 2-amino-2-cyclopropyl-2-(4-phosphonophenyl)acetic acid
Synonyms: amino(cyclopropyl)(4-phosphonophenyl)acetic acid
Molecular Weight (g/mol): 271.21
Molecular Formula: C11H14NO5P
InChi Key: IGODGTDUQSMDQU-UHFFFAOYNA-N
SMILES: NC(C1CC1)(C(O)=O)C1=CC=C(C=C1)P(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)