accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNNF

CAS RN 1835-65-0

IUPAC Name: tetrafluorobenzene-1,2-dicarbonitrile
Synonyms: tetrafluorophthalonitrile
Molecular Weight (g/mol): 200.1
Molecular Formula: C8F4N2
InChi Key: OFLRJMBSWDXSPG-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C#N)C(C#N)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)