accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaOOHNNOSOOOSOOO(Z)

CAS RN 1836-22-2

IUPAC Name: trisodium 2-{2-[(1Z)-2-oxo-3,6-disulfonato-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}benzoate
Synonyms: trisodium 2-{2-[(1Z)-2-oxo-3,6-disulfonatonaphthalen-1-ylidene]hydrazin-1-yl}benzoate
Molecular Weight (g/mol): 518.35
Molecular Formula: C17H9N2Na3O9S2
InChi Key: SJYYQLRKEKKFSX-NETMGOMMSA-K
SMILES: [Na+].[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1N\N=C1/C(=O)C(=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results