accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHCH3NH3CCH3

CAS RN 1836-42-6

IUPAC Name: benzyltriethylazanium hydroxide
Synonyms: benzyltriethylazanium hydroxide
Molecular Weight (g/mol): 209.33
Molecular Formula: C13H23NO
InChi Key: FKPSBYZGRQJIMO-UHFFFAOYSA-M
SMILES: [OH-].CC[N+](CC)(CC)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results