Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltriethylammonium Hydroxide (10% in Water), TCI America™
SDP

Catalog No. B044525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B044525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B044525ML Supplier TCI America Supplier No. B044525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1836-42-6
Molecular Formula C13H23NO
Molecular Weight (g/mol) 209.333
MDL Number MFCD00040488
InChI Key FKPSBYZGRQJIMO-UHFFFAOYSA-M
PubChem CID 74601
IUPAC Name benzyl(triethyl)azanium;hydroxide
SMILES CC[N+](CC)(CC)CC1=CC=CC=C1.[OH-]

Specifications

Color Colorless
Quantity 25 mL
UN Number 3267
Formula Weight 209.33
Physical Form Liquid
Chemical Name or Material Benzyltriethylammonium Hydroxide (10% in Water)
EINECSNumber (3)-2694
RTECSNumber BS9625000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.