accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 183610-70-0

IUPAC Name: 3-(trifluoromethyl)pyridin-2-amine
Synonyms: 3-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 162.12
Molecular Formula: C6H5F3N2
InChi Key: YWOWJQMFMXHLQD-UHFFFAOYSA-N
SMILES: NC1=C(C=CC=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results