accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONCH3NH3CCH3NNOHNOHOOCH3OH3C(?)

CAS RN 184162-64-9

IUPAC Name: 2-[5-(2,6-dimethoxyphenyl)-1-(4-{[3-(dimethylamino)propyl](methyl)carbamoyl}-2-(propan-2-yl)phenyl)-1H-pyrazole-3-amido]adamantane-2-carboxylic acid
Synonyms: 2-[5-(2,6-dimethoxyphenyl)-1-(4-{[3-(dimethylamino)propyl](methyl)carbamoyl}-2-isopropylphenyl)pyrazole-3-amido]adamantane-2-carboxylic acid
Molecular Weight (g/mol): 685.87
Molecular Formula: C39H51N5O6
InChi Key: LWULHXVBLMWCHO-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1C1=CC(=NN1C1=C(C=C(C=C1)C(=O)N(C)CCCN(C)C)C(C)C)C(=O)NC1(C2CC3CC(C2)CC1C3)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)