CAS RN 18435-45-5
IUPAC Name:
nonadec-1-ene
Synonyms:
1-nonadecene
Molecular Weight (g/mol):
266.51
Molecular Formula:
C19H38
InChi Key:
NHLUYCJZUXOUBX-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCC=C
1
–
1
of
1
results
1
1-Nonadecene 99.0+%, TCI America™
CAS: 18435-45-5 Molecular Formula: C19H38 Molecular Weight (g/mol): 266.51 MDL Number: MFCD00009010 InChI Key: NHLUYCJZUXOUBX-UHFFFAOYSA-N PubChem CID: 29075 ChEBI: CHEBI:64503 IUPAC Name: nonadec-1-ene SMILES: CCCCCCCCCCCCCCCCCC=C