accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNHNFClONO

CAS RN 184475-35-2

IUPAC Name: N-(3-chloro-4-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]quinazolin-4-amine
Synonyms: gefitinib tarceva gefitinib preparation iressa C22H24ClFN4O3 gefitinib (substance) gefitinib (product) irressat
Molecular Weight (g/mol): 446.91
Molecular Formula: C22H24ClFN4O3
InChi Key: XGALLCVXEZPNRQ-UHFFFAOYSA-N
SMILES: COC1=C(OCCCN2CCOCC2)C=C2C(NC3=CC=C(F)C(Cl)=C3)=NC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)