Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Iressa
GREENER_CHOICESDP

Catalog No. 300050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-005-0 50 mg
30-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-005-0 Supplier R&D Systems™ Supplier No. 3000/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Orally active, selective EGFR inhibitor

Iressa is an orally active, selective inhibitor of EGFR tyrosine kinase (IC50 = 23 - 79 nM). Shows minimal activity against ErbB2, KDR, c-flt, PKC, MEK and ERK-2. Blocks EGFR autophosphorylation and inhibits tumor growth in mice bearing a range of human xenografts. Also inhibits growth of brain metastases in mouse model of non-small cell lung cancer. View information regarding the usage of Iressa for non-clinical studies .

Chemical Identifiers

CAS 184475-35-2
Molecular Formula C22H24ClFN4O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 446.9
Purity >98%
Molecular Weight (g/mol) 446.91
Chemical Name or Material N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-morpholinyl)propoxy]-4-quinazolinamine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.