accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 184674-99-5

NOFOH2NHCl

CAS RN 184674-99-5

IUPAC Name: 3-(dicyclobutylamino)-8-fluoro-3,4-dihydro-2H-1-benzopyran-5-carboxamide hydrochloride
Synonyms: 3-(dicyclobutylamino)-8-fluoro-3,4-dihydro-2H-1-benzopyran-5-carboxamide hydrochloride
Molecular Weight (g/mol): 354.85
Molecular Formula: C18H24ClFN2O2
InChi Key: GSZJANKLCPHEEX-UHFFFAOYNA-N
SMILES: Cl.NC(=O)C1=C2CC(COC2=C(F)C=C1)N(C1CCC1)C1CCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results