accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClCl

CAS RN 18495-30-2

IUPAC Name: 1,1,2,3-tetrachloropropane
Synonyms: 1,1,2,3-tetrachloropropane
Molecular Weight (g/mol): 181.87
Molecular Formula: C3H4Cl4
InChi Key: BUQMVYQMVLAYRU-UHFFFAOYNA-N
SMILES: ClCC(Cl)C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results