accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 185017-72-5

IUPAC Name: 3-bromo-2-chloro-6-methylpyridine
Synonyms: 3-bromo-2-chloro-6-methylpyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: JVDQYSIJBRTRMS-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C(Cl)=N1
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)