accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOHNO

CAS RN 1858242-02-0

IUPAC Name: 2-(7-benzoyl-3-oxo-3,4-dihydroquinoxalin-2-yl)-2-phenylacetonitrile
Synonyms: 2-(7-benzoyl-3-oxo-4H-quinoxalin-2-yl)-2-phenylacetonitrile
Molecular Weight (g/mol): 365.39
Molecular Formula: C23H15N3O2
InChi Key: JAHIUCLUJXQZRA-UHFFFAOYNA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C2NC(=O)C(=NC2=C1)C(C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results