accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHSH2NNO(E/Z)

CAS RN 1858264-29-5

IUPAC Name: 1-[2-(1,4-dihydropyridin-4-ylidene)-4-phenyl-2,5-dihydro-1,3-thiazol-5-ylidene]-1-nitrosomethanamine
Synonyms: 1-nitroso-1-[4-phenyl-2-(1H-pyridin-4-ylidene)-1,3-thiazol-5-ylidene]methanamine
Molecular Weight (g/mol): 296.35
Molecular Formula: C15H12N4OS
InChi Key: VKJURGBMUDKEDZ-UHFFFAOYSA-N
SMILES: NC(N=O)=C1SC(N=C1C1=CC=CC=C1)=C1C=CNC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results