accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186392-43-8

IUPAC Name: 3-[(5-chloro-1H-indol-2-yl)formamido]-2-hydroxy-N-methoxy-N-methyl-4-phenylbutanamide
Synonyms: 3-[(5-chloro-1H-indol-2-yl)formamido]-2-hydroxy-N-methoxy-N-methyl-4-phenylbutanamide
Molecular Weight (g/mol): 415.87
Molecular Formula: C21H22ClN3O4
InChi Key: WEQLRDLTRCEUHG-UHFFFAOYNA-N
SMILES: CON(C)C(=O)C(O)C(CC1=CC=CC=C1)NC(=O)C1=CC2=CC(Cl)=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)