accessibility menu, dialog, popup

UserName

Viewing profile as user:
IOClF

CAS RN 186584-73-6

IUPAC Name: 2-fluoro-5-iodobenzoyl chloride
Synonyms: 2-fluoro-5-iodobenzoyl chloride
Molecular Weight (g/mol): 284.45
Molecular Formula: C7H3ClFIO
InChi Key: CEMYZMDITJKYDA-UHFFFAOYSA-N
SMILES: FC1=C(C=C(I)C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results