Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-iodobenzoyl chloride, 98%, Thermo Scientific™

Catalog No. AAH2617703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2617703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2617703 Supplier Thermo Scientific Chemicals Supplier No. H2617703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186584-73-6
Molecular Formula C7H3ClFIO
Molecular Weight (g/mol) 284.453
MDL Number MFCD03094145
InChI Key CEMYZMDITJKYDA-UHFFFAOYSA-N
PubChem CID 2778204
IUPAC Name 2-fluoro-5-iodobenzoyl chloride
SMILES C1=CC(=C(C=C1I)C(=O)Cl)F

Specifications

Melting Point 27°C to 30°C
Flash Point 59°C (139°F)
Quantity 1 g
UN Number UN3261
Sensitivity Light and moisture sensitive
Synonym benzoyl chloride,2-fluoro-5-iodo, acmc-20anuw, pubchem2131, 2-fluoro-5-iodobenzoylchloride, benzoyl chloride, 2-fluoro-5-iodo, 2-fluoranyl-5-iodanyl-benzoyl chloride, 2-fluoro-5-iodobenzoyl chloride
Formula Weight 284.45
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-iodobenzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.