accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SOONHNOHOHNOOCH3CH3H3CNH2CH3OCH3H3C(E/Z)

CAS RN 186698-61-3

IUPAC Name: 5-{[amino(2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-sulfonamido)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[amino(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-sulfonamido)methylidene]amino}-2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 526.65
Molecular Formula: C24H38N4O7S
InChi Key: CVFXPOKENLGCID-UHFFFAOYNA-N
SMILES: CC1=C2CC(C)(C)OC2=C(C)C(C)=C1S(=O)(=O)NC(N)=NCCCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)