accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CONHNFONOHO

CAS RN 186826-86-8

IUPAC Name: hydrogen 1-cyclopropyl-6-fluoro-8-methoxy-7-{octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl}-4-oxo-1,4-dihydroquinoline-3-carboxylic acid chloride
Synonyms: hydrogen moxifloxacin chloride
Molecular Weight (g/mol): 437.9
Molecular Formula: C21H25ClFN3O4
InChi Key: IDIIJJHBXUESQI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CC2CCCNC2C1)C1CC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (3)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)