Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moxifloxacin HCl, G-Biosciences
SCI_ED

Catalog No. S000002
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000002 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000002 Supplier G-Biosciences Supplier No. RC1058
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moxifloxacin is a broad-spectrum fourth generation fluoroquinolone antibiotic. It targets both gram-negative and gram-positive bacteria.

It has been shown to combat methicillin-susceptible Staphylococci species as well as penicillin-resistant and penicillin-susceptible Pneumococci and Streptococci species.

Chemical Identifiers

CAS 186826-86-8
Molecular Weight (g/mol) 437.89 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 500 mg
Percent Purity >98%
Grade Molecular Biology
Chemical Name or Material Moxifloxacin HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.