accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18691-97-9

IUPAC Name: 1-(1,3-benzothiazol-2-yl)-1,3-dimethylurea
Synonyms: methabenzthiazuron
Molecular Weight (g/mol): 221.28
Molecular Formula: C10H11N3OS
InChi Key: RRVIAQKBTUQODI-UHFFFAOYSA-N
SMILES: CNC(=O)N(C)C1=NC2=CC=CC=C2S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results