accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18695-01-7

IUPAC Name: 4-{4,5-dihydroxy-9-methyl-3-oxo-1H,3H-naphtho[2,3-c]furan-1-yl}-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide
Synonyms: 4-{4,5-dihydroxy-9-methyl-3-oxo-1H-naphtho[2,3-c]furan-1-yl}-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide
Molecular Weight (g/mol): 442.42
Molecular Formula: C22H22N2O8
InChi Key: DRKMHDAKULCOKQ-UHFFFAOYNA-N
SMILES: CN(C)C1C(C2OC(=O)C3=C2C(C)=C2C=CC=C(O)C2=C3O)C(O)C(=O)C(C(N)=O)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)